Learn More
United Scientific™ Double Walled Calorimeter
High quality and durable double walled calorimeter.
Supplier: United Scientific™ CLRM01
This item is not returnable.
View return policy
Description
- This calorimeter consists of two nested aluminum cans separated by a 1.7 cm thick styrofoam liner and a clear molded plastic lid with a filler cap
- A plastic insulator ring protects the styrofoam insulation against damage and liquid spills
- The inner reservoir has a capacity of 150 mL
- A thermometer stopper and an insulated stirrer are included
- Includes activity guide
Specifications
Calorimeter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Prop 65