Learn More
Thermo Scientific™ Double Bagged Bottled Tryptone Soya Broth (TSB)
Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.
Supplier: Thermo Scientific™ R112751
Description
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Tryptone Soya Broth (TSB) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Tryptone Soya Broth (TSB) is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Add Remel Double Bagged Bottled Tryptone Soya Broth (TSB) to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.Complete Solutions, Complete Confidence:• Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media• Regulatory compliant: USP, JP, and EU harmonized media • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs• Quality guaranteed: Certificates of Analysis with every lot produced • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle• Multi-purpose media: suitable for general quality testing Customizable formats and media types are available. Please inquire.A highly nutritious versatile medium is recommended for general laboratory use. Due to the inclusion of both Tryptone and Soya Peptone, the medium will support a luxuriant growth of many fastidious organisms without the addition of serum.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.Specifications
Stopper, Crimp seal | |
2 to 25°C | |
Double Bagged Serum Bottle | |
100 mL |
Stopper, Crimp seal | |
Double Bagged Bottled Tryptone Soya Broth (TSB) | |
Microbiology Media |
Safety and Handling
ShelfLife : 365 Days
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.