Learn More
Solutions Inc DOLLY FOR WASTE BOXES
Supplier: Solutions Inc 3804
4 Wheel Dolly, White for Corrugated Box. Fold down flaps; Fit hinged lid over flaps; Place empty box on Dolly; Remove full box Reuse lid & dolly; Replace with an empty box