Learn More
Enzo Life Sciences Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid (10mg). CAS: 24880-45-3
Supplier: Enzo Life Sciences BMLFA0120010
Polyunsaturated fatty acid. Purity: ≥98%. Solubility: Soluble in ethanol, hexane and methylene chloride. Long Term Storage: -20°C.