Learn More
Cole-Parmer Amprobe 33XR DMM, Prof TRMS with Temp, Backlit
Supplier: Cole-Parmer 2000548
True RMS digital multimeter with temperature and backlight has a frequency up to 40 MHz and temperature up to 1832 F. It also features the following: capacitance up to 4000 F; 1000 V/750 V dc/ac; 10 amps ac/dc; fully safety fused; resistance to 40 M; continuity beeper, and input warning tone when test leads are in wrong jack.