Learn More
Cole-Parmer Amprobe 30XR DMM, Manual Ranging, NC voltage (all ranges: +/-(1.5 % rdg + 4 dgts))
Supplier: Cole-Parmer 2000545
This non-contact voltage tester has a maximum of 600 AC volts, and 10 amps; resistance to 20 M; continuity beeper; 1.5 V / 9 V battery test under load and comes with Magne-Grip™ Holster with magnetic hanging strap.