Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva DMEM, Powder with Low Glucose
Supports cell culture growth and biomanufacturing processes with powdered DMEM glucose media. HyClone™ DMEM, Powder with Low Glucose is manufactured using ISO 9001 processes.
Supplier: Cytiva SH30002.04
This item is not returnable.
View return policy
Description
- High-quality classical media formulations in convenient packaging
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
- Exacting manufacturing processes ensure lot–to–lot consistency
Specifications
cGMP and ISO 9001:2000 | |
Powder | |
Non-sterile | |
Glutamine, Glucose, Sodium Pyruvate, Phenol Red, Sodium Bicarbonate |
2° to 8°C, away from light and moisture | |
DMEM/Low Glucose with L-glutamine, sodium pyruvate | |
Non-sterile | |
50 L |
Safety and Handling
ShelfLife : 36 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction