Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Adjusts the unit from 28 to 40 in.H with the push of a button
- Plenty of workspace
- Top made up of solid phenolic resin or epoxy resin which resists water, heat and most chemicals
- Two hooks mounted under the worksurface holds up to four book bags
- Bone powder-coated steel and oak veneer base conceals the lifting mechanism
- Motor is accessible by removing one of the oak panels
- Provides six GFI protected electrical outlets
- Easy, power option
Compliance
Meets ADA specifications
Warranty and Services
Limited lifetime
Specifications
Power-lift Workstation | |
243.84 cm | |
96 in. | |
127 cm | |
28 to 40 in. | |
299.37 kg |
660 lb. | |
71.1 to 101.6 cm | |
Ideal for lab | |
Chemical Resistant UV | |
50 in. | |
Wood |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction