Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Oak Table with Plain Aprons
Outfit your lab with easy-to-maintain furniture. Diversified Oak Table with Plain Aprons are made of smoothly machined oak, providing long-lasting usability and easy care.
Supplier: Diversified Spaces™ P7102K30N
Description
Solid red oak tables with plain aprons and a chemical-resistant finish. Fitted with 2.25 in. (5.7cm) square oak legs.
- Three tops available: plastic laminate, ChemGuard™ and 1 in. (2.54cm) epoxy resin
- Legs installed with 0.31 in. (0.79cm) hanger bolts and attached to apron with supplied nuts and washers
- Legs equipped with nonskid adjustable glides for precise leveling
- Legs fashioned with rubber leg boots
- Apron fitted with heavy-duty 13-gauge steel corner braces
Specifications
Table | |
48 in. | |
48 x 24 x 30 in. (121.92 x 60.96 x 76.2 cm) | |
ChemGuard™ | |
76.2 cm | |
121.92 cm | |
24 in. | |
Oak |
60.96 cm | |
30 in. | |
Plain apron, Steel corner braces | |
70 lb. | |
60.96 cm | |
31.2 kg | |
24 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction