Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Eye Safety Cabinet

Monitor provides excellent protection in five minutes and automatically shuts off if the double doors are opened.
Supplier: Diversified Spaces™ ESC1111LM
Description
- 24-gauge white enameled steel
- Pilot light confirms UV lamp is in use
- Five-minute cycle controlled by timer
- Doors interlocked with tamper-resistant latches
- Wall and shelf mountable with 7 ft. (2.1m) grounded electrical cord
Specifications
White | |
9.5 in. | |
1 Ea. | |
32 in. |
6 pairs of glasses or 5 pairs goggles per shelf on 8 shelves | |
Eye Safety Cabinet | |
Steel | |
24.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title