Learn More
Tecan 100mL Disposable Trough
Supplier: Tecan 10613049
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Made of gray polypropylene and is autoclavable. Dead volume approximately 3. 3 mL. Scaling on trough every 10 mL from 10 - 100 mL. For use in Tecan carrier p/n 10613020. 18 bags of 6 - 108 troughs/sales unit.
This item is not returnable.
View return policy