Learn More
Braintree Scientific Disposable Safety Scalpel, S/S #22, 10/pkg
Supplier: Braintree Scientific 4522
Disposable Stainless Steel Safety Scalpel - Engineered for safety and simplicity. Complies with the OSHA directive by providing a fully retractable scalpel blade, which protects the user from accidental needle sticks associated with traditional scalpels. Packaged 10 Safety Scalpels per dispenser box, sterile, single use. Reusable sterile stainless dissecting surgical blades and handles also available - call for size and pricing. **Sold for research purposes only and not for human use**
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.