Learn More
Valutek™ Disposable Polypropylene Bouffant Cap
Click to view available options
Size: 21 in.
Description
- Filament covered unexposed elastic edge design for secure fit and unexposed closure
- All Valutek™ bouffant caps are tested and manufactured in ISO compliant facilities under individual inspection and strict process
- Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environments
Specifications
Disposable | Single-use |
Model | VTBFCW-21 |
Cleanroom Class | Class 10,000 (ISO 7) |