Learn More
Fisherbrand™ Disposable Inoculating Loops
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
Description
- Ideal for anaerobic microbiology applications where deep agar inoculation is required
- Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
- High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
- Rigid enough to penetrate the media for subsurface inoculation
- Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
- Can be used under hoods without danger
- Color-coded for ease of size identification
Specifications
Yes | |
Polystyrene | |
Blue | |
No |
10 μL | |
Non-sterile | |
20 Loops |