Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Disposable Face Shield
Click to view available options
With Anti-Fog Coating
Description
Now more than ever, PPE has become essential in the fight to address the growing risk of exposure to splash, droplet and airborne pathogens we cannot see.
The disposable face shield complies with Government regulations for splash and droplets, and offers enhanced protection against the transmission of droplets in the air. Unlike DIY solutions, this face shield provides an antifog coating to reduce moisture build-up and avoid low visibility and hazing.
Features and Benifits
- Provides an extra layer of protection when social distancing isn't possible
- 15 in. wide and 9.1 in. height to provide large coverage over the face
- PET recyclable material with 0.188 mm thickness
- Anti-fog coating on interior surface to prevent fogging and provide better vision
- 1.2 in. foam thickness for optimal distance between face and face shield
- FDA registered
- Complies with: ANSI/ISEA Z87.1-2015 Clause 8.1.2 droplet and splash hazard-face shields, EN 166:2001 Clause 7.2.4 protection against droplets and splashes of liquids
- Elastic band (0.6 in. wide) for extra comfort and universal fit
- White background for easy writing names or functions
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Width (English) | 15 in. |
Height (English) | 9.1 in. |
Thickness (Metric) | 0.188 mm |
Lens Coating | Anti-fog |
Lens Tint | Clear |
For Use With (Application) | For use in the following industries: Medical personnel, cashiers, store employees, banks, government offices, restaurent staff |
Headband Material | Foam |
Quantity | 200/Pk. |
Material | PET |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction