Learn More
Andwin Scientific DISPOSABLE CULTURE TUBES DURE
Supplier: Andwin Scientific 47729566
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef