Learn More
Andwin Scientific DISPOSABLE CULTURE TUBE
Supplier: Andwin Scientific 47729576
Culture Tube Disposable, Borosilicate Glass, Chemically inert. High heat tolerance. Uniform dimensions. Clean-sealed in a convenient dispensing package. 16 O.D. x 100L mm, 14mL overflow Volume,