Learn More
Robbins Instruments Inc Robbins True-Cut Disposable Biopsy Punch 0.50mm, 25/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Robbins Instruments Inc RBP05
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These devices, constructed from 403L stainless steel, consist of extremely sharp cutting edges. The punch has the capability to quickly cut and retrieve different types of tissue samples. One of the most important features of this device is that they are sterile and disposable.
This item is not returnable.
View return policy