Learn More
BRUKER BIOSPIN HR-MAS Disposable Insert Kit
Supplier: BRUKER BIOSPIN B4493
To be used with 4mm Bruker MAS rotors. Each Kit includes: 10 Kel-F insert, 10 Sealing Screws, 10 Plugs, 1 Forked Screwdriver, 1 Blunt-ended Screwdriver, 1 Extraction Screw.