Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Dispensing Trays
Dispensing valve at one corner, close to bottom.
Description
- Durable 0.25 in. thick (6.4 mm) high-density polyethylene tray
- Welded corners and bottom
Order Info
Additional dispensing tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
12 x 16 x 3 in. (30.5 x 40.5 x 7.5 cm) | |
30.5 cm | |
Dispensing Tray | |
12 in. | |
Rectangular | |
16 in. |
7.5 cm | |
HDPE | |
40.5 cm | |
3 in. | |
0.63 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction