Learn More
BrandTech™ BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Supplier: BrandTech™ BRAND™ 704282
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Dispenser Thread Adapter | |
Connecting remote dispensing system with drums with GL outer thread | |
1 Ea. |
PTFE | |
Dispensette™III, Dispensette™ Organic Remote Dispensing Unit | |
32 GL (outer thread) |