Learn More
Hach Company Dispenser, Variable Volume, Digital, 1.0-10.0 mL
Supplier: Hach Company 2563140
Attach dispenser directly to reagent bottle for adjustable dispensation with a single stroke. Inert hardware designed to handle alkaline, acidic and organic solutions. Autoclavable (121°C). Dispensers fits bottles with a 33 mm screw-neck finish (supplied with adapters to fit 28, 38 and 45 mm finishes). Dispenser rotates 360°.