Learn More
Acuderm Inc Acu-Punch; 12mm; Acuderm, 25/box
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Acuderm Inc P1225
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Acu-Punch; 12mm; Acuderm;; Sterile; Disposable Skin Biopsy Punch; Only punch available in a complete range of 14 sizes from 1 to 12mm; Razor sharp and ready to use; Handle is ribbed for maximum control even when wearing gloves; 25/BX. *Item is non returnable