Learn More
Bellco Glass, Inc. For use in the disinfectant testing procedure as described in aqac manual. Cylinder ends are lightly glazed. Hard surface carrier test for efficacy of testing of disinfectants. Sizes: 8x8mm
Supplier: Bellco Glass, Inc. 209100808
For use in the disinfectant testing procedure as described in aqac manual. Cylinder ends are lightly glazed. Hard surface carrier test for efficacy of testing of disinfectants. Sizes: 8x8mm