Learn More
Andwin Scientific DISCIDE ULTRA TOWELETTES
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Andwin Scientific 60DIS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The DisCide Ultra Disinfecting Towelettes provide the quickest possible disinfection times. This product is able to eliminate TB, HIV-1 (AIDS) virus, Herpes 2 and Andenovirus 2 in 1 minute. Its self-timing evaporation gives assurance of disinfection, leaving a light herbal scent.
This item is not returnable.
View return policy