Learn More
Azzota Corp Disc Polarimeter, Features Visual Aim, Manual Measure, Easy Operation, Measuring Range +/-180Deg., Dial Venire Value In Reading 0.05Deg., Accuracy +/-0.05Deg., Sensitivity 0.05Deg., Mono Sodium Lamp
Supplier:  Azzota Corp DISCPOLAR
Encompass_Preferred
Disc Polarimeter, Features visual aim, manual measure, easy operation, Measuring range +/-180deg., Dial Venire Value in Reading 0.05deg., Accuracy +/-0.05deg., Sensitivity 0.05deg., Monochromatic Sodium lamp
Catalog No. 50-753-2959