Learn More
Electron Microscopy Sciences Dip Miser Cup Replacement
Supplier: Electron Microscopy Sciences 70520
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Glass cup is Pyrex and is designed to conserve expensive autoradiographic emulsion.