Learn More
Electron Microscopy Sciences Dip Miser 1set
Supplier: Electron Microscopy Sciences 70510
Slide coating cup with stand and cleaning brush. Glass cup is Pyrex and designed to conserve expensive autoradiographic emulsion when coating glass slides. Needs only 1/4 the volume of solution previously required. Single piece holder permits immersion in a water bath and provides a stable darkroom stand to prevent spills, and faster temperature adjustment of the emulsion.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.