Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
Supplier: Thermo Scientific™ 088782
Description
- Unequalled high-resolution separations
- Exceptionally high efficiency
- Highest resolution of monoclonal antibody variants
- Unmatched column-to-column and lot-to-lot reproducibility
- Hydrophobic interactions eliminated
- Excellent for stability testing
Specifications
1 Lot of Resin | |
Bio-LC |