Learn More
Thermo Scientific CS16-4um and CS16-Fast-4um IC Columns, 4 mm X 250 mm
Supplier: Thermo Scientific 088584
Analyze disparate concentration ratios up to 10,000:1 of close-eluting cations such as ammonium and sodium using CS16-4um and CS16-Fast-4um columns.