Learn More
Thermo Scientific DIONEX CDRS600 SUPPRESSOR
Supplier:  Thermo Scientific 088668
Encompass
An electrochemical suppressor that removes ions and conductivity from chromatography eluents by electrolytically converting the eluent to water with a recirculating current, enhancing analyte response; designed for high pressure ion chromatography systems to suppress background conductivity from carbonate/bicarbonate eluents for sensitive detection.
Catalog No. NC2249906
Explore available promotions