Learn More
Electron Microscopy Sciences Digital Microscopes Model AF4915ZTL, Type: USB 2.0. Magnification: 10x - 140x., 1/EA
Supplier: Electron Microscopy Sciences AF4915ZTL
The Dino-Lite Edge MP is an all-in-one device that was designed to meet professional applications within 10x - 140x magnifications at longer working distance. It integrates premium Edge Series features Extended Depth of Field (EDOF). Which is an easy to use stacking feature that captures several images at different focal points and then stacks them automatically into one clear image. This Dino-Lite microscope model connects via USB, but can also be wireless with the optional WF-20 Wireless Streamer, adding further versatility. Thanks to a long battery and stable WiFi image transmission, the WF-20 makes it a a completely cordless and portable microscope that is well suited for field applications. You can also use the WF-20 as a wireless router for staying connected to the internet or network during use. 1.3 MP with Automatic Magnification Reading (AMR) and Flexible LED Control (FLC). Type: USB 2.0. Magnification: 10x - 140x .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.