Learn More
Dino Lite VGA Digital Microscope AM5216TF- 720p, 1x - 70x Optical Magnification, Far Working Distance
Supplier: Dino Lite AM5216TF
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The AM5216TF provides extra-long working distance with a magnification range of 1x - 70x and features a High Definition VGA connection and improved optics for sharper images. It easily allows for the observation of objects from further away with a minimum working distance of 115mm at 70x. Capable of 60 frames per second at 720p (1280 x 720), this model is ideal for observing specimens effectively with minimum lag while in motion. This model is suitable for a wide range of applications including the inspection, repair and assembly involving reflective or translucent materials, with examples such as soldering, quality inspection, or jewelry fabrication. No computer or software required for this model that connects directly to a monitor or TV with VGA connections.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.