Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl Sulfoxide, USP, 99.9%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Dimethyl Sulfoxide, USP is also known as DMSO. It is used topically to decrease pain and speed the healing of wounds, burns, and muscle and skeletal injuries. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Packaging | Drum |
Refractive Index | 1.4755-1.4775 |
Quantity | 200 L |
Specific Gravity | 1.095-1.101 |
Percent Purity | 99.9% |
Grade | USP |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction