Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide, 67-68-5, MFCD00002089, 5x10mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences D24385X10ML
Linear Formula (CH3)2SO, Molecular Weight 78.13, BioPerformance Certified, suitable for hybridoma, Ready-to-use sterile filtered product conveniently packaged in septum screw-capped amber bottles. Meets USP and EP testing requirements.