Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroergotamine Mesylate, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog Number 18602211
Click to view available options
Quantity:
250 mg
1 g
5 g
100 g
Packaging:
Amber Glass Bottle
6190-39-2
C34H41N5O8S
679.79
MFCD00058615
ADYPXRFPBQGGAH-UMYZUSPBSA-N
(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
6190-39-2
C34H41N5O8S
679.79
MFCD00058615
ADYPXRFPBQGGAH-UMYZUSPBSA-N
(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O

D1165, 6190-39-2, C33H37N5O5•CH4O3S

Spectrum™ Chemical Dihydroergotamine Mesylate, USP is an ergot alkaloid medication used to treat active migraine headaches. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 4%
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -1°6.7° to -22.7°
Percent Purity 97 to 103%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit