Learn More
Cole-Parmer DIGTL MULTIMETER,CAT III 600V
Supplier: Cole-Parmer 2003648
Fluke 107 Palm-Sized Digital Multimeter, CAT III 600V. Reliable, easy-to-use multimeter fits right your shirt pocketOnly measures 2-3/4" x 5-5/8" x 1-1/8" (7 x 14.2 x 2.8 cm)Measures AC/DC voltage, AC/DC current, resistance, capacitance, and more!Backlit display for easy viewing