Learn More
Electron Microscopy Sciences Visio-Tek™ Digital Wi-Fi/USB Polarized Microscope, 5x ~ 200x magnification
Supplier: Electron Microscopy Sciences 92102
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Visio-tek digital portable imaging solutions utilize both Wi-Fi and USB transmission and can be connected to iOS and Android smartphones or tablets via the dedicated Visio-tek App, as well as to Windows or Mac computers or laptops through a Wi-Fi/USB connection. Using theVisio-tek free software and application, you can capture, store pictures and video, and take measurements. Flexible, cost effective and user-friendly. Visio-tek microscopes allow Wi-Fi inspections anywhere an image magnification is required. The polarizer decreases glare resulting in a darker, sharper image, with increased contrast.
Magnification: 5x ~ 200x
Focus range: 8 ~ 150 mm
Light source: 8 White LEDs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.