Learn More
Fisherbrand™ Digital Vortex Mixer
Ideal for applications that demand repeatable results.
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Includes: Includes both cup head and 3 in. head with cover. Units are supplied with a 72 in. (183cm), 3-wire cord and plug.
Description
Microprocessor control maintains set point for strong, consistent mixing action. Timer will display elapsed time or, when programmed to user defined time limit, the unit will shut off when the time reaches zero. Two modes of operation: “Continuous” mode when using accessory attachments or “Touch” mode which activates mixing when depressing the cup head.
- LED displays for speed and time
- Continuous or touch modes
- Microprocessor controls
- 5 year warranty
Warranty and Services
5 years
Specifications
4.9 mm | |
230 V, 0.13 A, 135 W | |
135 W | |
Digital Vortex Mixer | |
12.7 cm | |
20.2 cm | |
12.1 cm | |
5.3 kg | |
4.5 kg | |
5 year limited warranty on parts and labor. |
500 to 3000 rpm | |
230 V | |
0.13 A | |
5 in. | |
8in. | |
4.75 in. | |
10 lb. | |
10 lb. | |
Includes both cup head and 3 in. head with cover. Units are supplied with a 72 in. (183cm), 3-wire cord and plug. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.