Learn More
JDM SCIENTIFIC LLC DIGITAL TUBE ROTATOR
Supplier: JDM SCIENTIFIC LLC TR04U
The title angle is adjustable for accommodating different applicationsVariable sizes of Tube Holders and Carousels are available to accommodate various sizes of TubesAll units are compatible with a cold room and incubator environmentTR-04UA and TR-05UA models have speed adjustment, continuous and timer mode with automatic stop