Learn More
HANNA INSTRUMENTS INC Digital PEI Body pH Electrode with Matching Pin for Field Applications
Supplier: HANNA INSTRUMENTS INC HI12301
The HI12301 is a plastic body, double junction, gel filled pH electrode with a matching pin and built-in temperature sensor for temperature compensated measurements in a single probe design. This probe features a 3.5 mm connector designed exclusively to be used with Hanna Instruments edge Tablet Meters. The HI12301 electrode features a built-in microchip that stores sensor type, serial number, and calibration information. It also features a single ceramic junction and the spherical pH sensing portion is made with general purpose glass. The electrode has a rugged PEI (polyetherimide) body that has the chemical resistance of a glass body in an unbreakable plastic making it ideal for field and industrial applications.Spherical Glass TipDouble Junction Reference3.5 mm Connector
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.