Learn More
Cole-Parmer Wika A-AI.1 Attachable Loop-Powered Digital Pressure Indicator
Supplier: Cole-Parmer 6844713
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Installs between transmitter and DIN 43 650, + full sized "L" shaped 4-pin connector. Programmable Range: 4-20 mA can be assigned any display value within the display range. User scalable to display range from -1999 to +9999. Accuracy: = 0.2% of span.