Learn More
Seca Corp 813 High capacity digital flat scale for individual patient use with Bluetooth connectivity feature
Supplier: Seca Corp SECA 813 BLUETOOTH
seca has designed the seca 813 with a high capacity of up to 440 lbs and an extra-wide platform. The tough rubber mat ensures a secure stance and flat construction allows for easy accessibility. • Solid steel frame, up to 440 lbs capacity • Non-slip rubber base for safe standing during measuring • Large LCD figures for easy reading • Bluetooth connectivity for connection to Google Fit or Apple Health (via OKOK International bridge app) • Step-off function for weighing without switching on scale. • Capacity 440 lbs / 200 kg • Graduation 0.2 lb / 100 g • Functions: Auto-HOLD, lbs/kg switch-over, Step-off, tap-on automatic switch-on, automatic switch-off
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.