Learn More
Thermo Scientific™ Digital Dry Baths/Block Heaters
Description
Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
- Digital controls and display of time and temperature
- Advanced internal temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
- Powder-coated steel body construction ensures durability
- Built-in over-temperature protection device warrants sample and user safety
- Temperature deviation adjustment
- Sample preparation
- Enzyme preservation
- Enzyme-substrate reactions
- DNA amplification
- Electrophoresis gel degeneration
- Serum coagulation
- Unit must be ordered with block(s)
Specifications
Certifications/Compliance | CE, cULus, RoHS |
Controller Type | PID digital |
Material | Powder-coated Steel |
Plug Type | CN, EU, UK |
Relative Humidity | <80% |
Temperature Range (Metric) | Ambient +5° to 130°C |
Type | Dry Bath/Block |
Fuse | 250V 2.5A |
Heating Rate | ≤ 20 min. 30° to 130°C |
Includes | Dry bath, CN, EU and UK plugs, block lifter |
Show More |
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.