Learn More
Seca Corp 769 Digital column scale with BMI function
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Seca Corp 7691321004
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Versatile, low-maintenance and site-independent with economic battery operation ‒ the seca 769 is ideal for daily use in hospitals and doctors offices. With its transport castors, the column scale is also easy to transport. The seca 769 is equipped with several intelligent functions. For example, the BMI function permits a reliable evaluation of the nutritional condition of the patient. With the slip resistant base and anti-tip column design, patient scan be safely measured and weighed. • Particularly robust and compact column scale • Includes BMI function, TARE and HOLD • Weighing platform made of steel • Easily movable with transport wheels • Batteries (included) or optional power adapter • Capacity 450 lbs / 200 kg • Graduation 0.2 lb / 100 g • Functions: • TARE, HOLD, BMI, lbs/kg switch-over, automatic switch-off • Measuring rod seca 220 (included)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.