Learn More
Seca Corp 777 Digital column scale with eye-level display, handrail-compatible
Supplier: Seca Corp 7771321004
The seca 777 digital column scale, handrail-compatible (optional seca 477 handrail accessory) with eye-level display, is the medical-grade solution for measuring and weighing. The oversized, low-profile full metal platform accommodates all patient types and the tilt-proof safety design ensures the scale will not tip over when placing all weight on the edge of the base. In addition, the built-in measuring rod has a 4 – 90 inches / 10 – 230 cm measuring range. • Built-in measuring rod with 4 – 90 inches / 10 – 230 cm measuring range • 8-year warranty included (4x industry standard) • Large rubber coated wheels for excellent movability • Oversized, low-profile full metal platform (no plastic) • Tilt-proof safety design • Capacity 550 lbs / 250 kg • Graduation 100 g • Functions: TARE, HOLD, lbs/kg switch-over lock, BMI • Integrated measuring rod (included) • Measuring range 10 – 230 cm • Graduation 1 mm • Accessories: Power adapter seca 401, Handrail seca 477
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.