Learn More
Techcyte Digital Cervical Cytology Analysis
Digital Cervical Cytology Analysis (Pap smear): by combining our algorithm’s ability to confirm proposed atypical cells and organisms, Techcyte’s solution increases detection accuracy while decreasing time and cost.
Supplier: Techcyte PAPPAYG
Description
- Assist in the screening of SurePath and ThinPrep slides, scanned on various whole slide image scanners
- Detects evidence of squamous carcinoma, adenocarcinoma, and their typical precursor conditions
- Identifies and presents to a trained cytotech the cells most likely to be abnormal, which are reviewed on monitor
- When atypica is identified using Techcyte, the slide is reviewed manually using a microscope
Specifications
Detection Software | |
Analyzing Data |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Techcyte is CE-IVDD marked in Europe, and for investigational use only in the US.