Learn More
Seca Corp 334 Mobile digital baby scale
SDP
Supplier:  Seca Corp 3341321008
Encompass_Preferred
The weighing tray has a generous and comfortable design for babies and toddlers. The easy-to-transport seca 334 weighs only 6.2 lbs and requires surprisingly little space. With the large, clear LCD figures and the practical carrying handle which can be used to hang the scale, makes daily weighing very easy. The Breast-Milk-Intake-Function shows the exact amount of milk the baby has consumed. • Breast-Milk-Intake-Function (BMIF) • Handy and light for mobile medical use • Comfortable cradle • Large, easy to read LCD display • Reliable results due to automatic damping system • Extendable with optional measuring rod seca 232 • Capacity: 44 lbs / 20 kg • Graduation: 0.2 oz < 22 lbs > 0.5 oz / 5 g < 10 kg > 10 g • Functions: TARE, BMIF, auto-HOLD, lbs/kg switch-over, automatic switch-off • Battery operated (included) • Optional: measuring rod seca 232, head positioner seca 419, power adapter seca 447, switch-mode power adapter seca 400, carrying case seca 428
Catalog No. NC1798765
Explore available promotions