Learn More
Azzota Corp Digital Stirring Heating Mantle, Capacity: 500Ml, 250W, Maximum Temperature Is 450'C. Stir Speed: 0 To 1400Rpm.
Supplier: Azzota Corp DSHM500
Digital Stirring Heating Mantle, Capacity: 500ml, 250W, Maximum temperature is 450'C. Stir speed: 0 to 1400rpm.