Learn More
Azzota Corp Digital Stirring Heating Mantle, Capacity: 2000Ml, 350W, Maximum Temperature Is 450'C. Stir Speed: 0 To 1400Rpm.
Supplier: Azzota Corp DSHM2000
Digital Stirring Heating Mantle, Capacity: 2000ml, 350W, Maximum temperature is 450'C. Stir speed: 0 to 1400rpm.